Anti-APOA4 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB28585EA 568 EUR
ABNOPAB28585
Anti-APOA4 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant APOA4.

Recommended Dilutions: Western Blot: 1:100-1:250; Immunofluorescence: 1-4 ?g/ml; Immunohistochemistry: 1:200-1:500

Type: Primary
Antigen: APOA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR