Anti-TTR Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB28586EA 568 EUR
ABNOPAB28586
Anti-TTR Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant TTR.

  • Recommended Dilutions: Western Blot: 1:100-1:250; Immunohistochemistry: 1:500-1:1000

Type: Primary
Antigen: TTR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR