Anti-CDG1S, CXorf45, GLT28D1, MDS031, TDRD13, YGL047W Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB28600EA 568 EUR
ABNOPAB28600
Anti-CDG1S, CXorf45, GLT28D1, MDS031, TDRD13, YGL047W Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant ALG13.

Recommended Dilutions: Western Blot: 1:100-1:250; Immunohistochemistry: 1:200-1:500

Type: Primary
Antigen: CDG1S, CXorf45, GLT28D1, MDS031, TDRD13, YGL047W
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR