Anti-RSHL1 Mouse Monoclonal Antibody [clone: 3D12]

Lieferant: Abnova
ABNOH00081492-M02EA 382 EUR
ABNOH00081492-M02
Anti-RSHL1 Mouse Monoclonal Antibody [clone: 3D12]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant RSHL1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: RSHL1
Clonality: Monoclonal
Clone: 3D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR