Anti-COL26A1 Mouse Monoclonal Antibody [clone: 2D10]

Lieferant: Abnova
ABNOH00136227-M01EA 394 EUR
ABNOH00136227-M01
Anti-COL26A1 Mouse Monoclonal Antibody [clone: 2D10]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant EMID2.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: COL26A1
Clonality: Monoclonal
Clone: 2D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR