Anti-PIK3R6 Mouse Monoclonal Antibody [clone: 1G16]

Lieferant: Abnova
ABNOH00146850-M01EA 382 EUR
ABNOH00146850-M01
Anti-PIK3R6 Mouse Monoclonal Antibody [clone: 1G16]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant PIK3R6.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: PIK3R6
Clonality: Monoclonal
Clone: 1G16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR