Anti-CNTLN Mouse Monoclonal Antibody [clone: 1A10]

Lieferant: Abnova
ABNOH00054875-M03EA 382 EUR
ABNOH00054875-M03
Anti-CNTLN Mouse Monoclonal Antibody [clone: 1A10]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant CNTLN.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: CNTLN
Clonality: Monoclonal
Clone: 1A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR