Anti-FHL3 Mouse Polyclonal Antibody

Lieferant: Abnova
ABNOH00002275-B02PEA 394 EUR
ABNOH00002275-B02P
Anti-FHL3 Mouse Polyclonal Antibody
Antikörper
Mouse polyclonal antibody raised against a full-length human FHL3 protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: FHL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR