Anti-CSTB Mouse Monoclonal Antibody [clone: S3]

Lieferant: Abnova
ABNOH00001476-M02AEA 382 EUR
ABNOH00001476-M02A
Anti-CSTB Mouse Monoclonal Antibody [clone: S3]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant CSTB.

Type: Primary
Antigen: CSTB
Clonality: Monoclonal
Clone: S3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR