Anti-HSPD1 Mouse Monoclonal Antibody [clone: 2A7]

Lieferant: Abnova
ABNOH00003329-M01EA 382 EUR
ABNOH00003329-M01
Anti-HSPD1 Mouse Monoclonal Antibody [clone: 2A7]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant HSPD1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: HSPD1
Clonality: Monoclonal
Clone: 2A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR