Anti-IGFBP3 Mouse Monoclonal Antibody [clone: 2G6]
Lieferant: Abnova
ABNOH00003486-M02EA 394 EUR
ABNOH00003486-M02
Anti-IGFBP3 Mouse Monoclonal Antibody [clone: 2G6]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant IGFBP3.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: IGFBP3
Clonality: Monoclonal
Clone: 2G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Bestellen