Anti-TADA2L Mouse Monoclonal Antibody [clone: S1]

Lieferant: Abnova
ABNOH00006871-M01AEA 394 EUR
ABNOH00006871-M01A
Anti-TADA2L Mouse Monoclonal Antibody [clone: S1]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant TADA2L.

Type: Primary
Antigen: TADA2L
Clonality: Monoclonal
Clone: S1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR