Anti-RCVRN Mouse Monoclonal Antibody [clone: 1H5]

Lieferant: Abnova
ABNOH00005957-M16CEA 382 EUR
ABNOH00005957-M16C
Anti-RCVRN Mouse Monoclonal Antibody [clone: 1H5]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant RCVRN.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: RCVRN
Clonality: Monoclonal
Clone: 1H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR