Anti-LMO2 Mouse Monoclonal Antibody [clone: 4D3]
Lieferant: Abnova
ABNOH00004005-M05CEA 394 EUR
ABNOH00004005-M05C
Anti-LMO2 Mouse Monoclonal Antibody [clone: 4D3]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant LMO2.
Recommended Dilutions: Western Blot: 1:500-1:1000
Type: Primary
Antigen: LMO2
Clonality: Monoclonal
Clone: 4D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human