Anti-LGALS7 Mouse Monoclonal Antibody [clone: 6B5]
Lieferant: Abnova
ABNOH00003963-M01EA 394 EUR
ABNOH00003963-M01
Anti-LGALS7 Mouse Monoclonal Antibody [clone: 6B5]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant LGALS7.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: LGALS7
Clonality: Monoclonal
Clone: 6B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Bestellen