Anti-NFYC Mouse Monoclonal Antibody [clone: 1D3]

Lieferant: Abnova
ABNOH00004802-M01AEA 382 EUR
ABNOH00004802-M01A
Anti-NFYC Mouse Monoclonal Antibody [clone: 1D3]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant NFYC.

Type: Primary
Antigen: NFYC
Clonality: Monoclonal
Clone: 1D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR