Anti-OPA1 Mouse Monoclonal Antibody [clone: 8A20]

Lieferant: Abnova
ABNOH00004976-M02EA 382 EUR
ABNOH00004976-M02
Anti-OPA1 Mouse Monoclonal Antibody [clone: 8A20]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant OPA1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: OPA1
Clonality: Monoclonal
Clone: 8A20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR