Anti-PFDN4 Mouse Polyclonal Antibody

Lieferant: Abnova
ABNOH00005203-B03EA 394 EUR
ABNOH00005203-B03
Anti-PFDN4 Mouse Polyclonal Antibody
Antikörper
Mouse polyclonal antibody raised against a full-length human PFDN4 protein.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: PFDN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR