Anti-IL17A Mouse Monoclonal Antibody [clone: 3G5]

Lieferant: Abnova
ABNOH00003605-M01EA 382 EUR
ABNOH00003605-M01
Anti-IL17A Mouse Monoclonal Antibody [clone: 3G5]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant IL17A.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: IL17A
Clonality: Monoclonal
Clone: 3G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR