Anti-MUC2 Mouse Monoclonal Antibody [clone: CCP58]

Lieferant: Abnova
ABNOMAB11323EA 454 EUR
ABNOMAB11323
Anti-MUC2 Mouse Monoclonal Antibody [clone: CCP58]
Antikörper
Mouse monoclonal antibody raised against MUC2.

Recommended Dilutions: Western Blot: 1:100-1:2000; Flow Cytometry: 1:10 - 1:1000; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:200 - 1:400; Immunoprecipitation: 1:10-1:500

Type: Primary
Antigen: MUC2
Clonality: Monoclonal
Clone: CCP58
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR