Anti-TP53 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: Pab 1801]

Lieferant: Abnova
ABNOMAB11420EA 515 EUR
ABNOMAB11420
Anti-TP53 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: Pab 1801]
Mouse monoclonal antibody raised against recombinant partial TP53.

Recommended Dilutions: Western Blot: 1:1000-1:2000; Immunohistochemistry: 1:100-1:250; Immunoprecipitation: 10 ?g/mg)

Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: Pab 1801
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR