Anti-DMPO Monoclonal Antibody (ATTO 700) [clone: N1664A]

Lieferant: Abnova
ABNOMAB11434EA 568 EUR
ABNOMAB11434
Anti-DMPO Monoclonal Antibody (ATTO 700) [clone: N1664A]
Mouse monoclonal antibody raised against DMPO.

Recommended Dilutions: ELISA: 1-10 ?g/ml; Immunocytochemistry: 10 ?g/ml; Immunoprecipitation: 25 ?g

Type: Primary
Antigen: DMPO
Clonality: Monoclonal
Clone: N1664A
Conjugation: ATTO 700
Epitope:
Host:
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR