Anti-VIM Mouse Monoclonal Antibody [clone: LN6]

Lieferant: Abnova
ABNOMAB11483EA 518 EUR
ABNOMAB11483
Anti-VIM Mouse Monoclonal Antibody [clone: LN6]
Antikörper
mouse monoclonal antibody raised against VIM.

Recommended Dilutions: Immunofluorescence: 1:50-1:100; Immunohistochemistry: 1:100-1:200 for 30 min at RT; Western Blot: 1:400-1:800

Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: LN6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR