Anti-ACTA1 Mouse Monoclonal Antibody [clone: HHF35]

Lieferant: Abnova
ABNOMAB11331EA 518 EUR
ABNOMAB11331
Anti-ACTA1 Mouse Monoclonal Antibody [clone: HHF35]
Antikörper
Mouse monoclonal antibody raised against muscle-specific ACTA1.

Recommended Dilutions: Flow Cytometry: 0.5-1 ?g/million cells; Immunofluorescence: 0.5-1 ?g/ml; Immunohistochemistry: 0.25-0.5 ?g/ml for 30 min at RT; Immunoprecipitation: 0.5-1 ?g/500 ?g protein lysate; Western Blot: 0.25-0.5 ?g/ml

Type: Primary
Antigen: ACTA1
Clonality: Monoclonal
Clone: HHF35
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR