Anti-CEACAM5 Mouse Monoclonal Antibody [clone: CEA31]

Lieferant: Abnova
ABNOMAB11336EA 518 EUR
ABNOMAB11336
Anti-CEACAM5 Mouse Monoclonal Antibody [clone: CEA31]
Antikörper
Mouse monoclonal antibody raised against CEACAM5.

Recommended Dilutions: ELISA Flow Cytometry: 0.5-1 ?g/million cells; Immunofluorescence: 1-2 ?g/ml; Immunohistochemistry: 1-2 ?g/ml for 30 min at RT; Immunoprecipitation: 1-2 ?g/500 ?g protein; Western Blot: 0.25-0.5 ?g/ml

Type: Primary
Antigen: CEACAM5
Clonality: Monoclonal
Clone: CEA31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR