Anti-Y Monoclonal Antibody [clone: PY20]

Lieferant: Abnova
ABNOMAB11325EA 518 EUR
ABNOMAB11325
Anti-Y Monoclonal Antibody [clone: PY20]
Antikörper
Mouse monoclonal antibody raised against Phosphotyrosine.

Recommended Dilutions: Western Blot: 1:200 -1:2000; Immunocytochemistry/Immunofluorescence: 1:10 - 1:2000; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:50-1:100; Immunoprecipitation: 1:10 - 1:500

Type: Primary
Antigen: Y
Clonality: Monoclonal
Clone: PY20
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2b kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR