Anti-KRT1 Mouse Monoclonal Antibody [clone: AE1 + AE3]

Lieferant: Abnova
ABNOMAB11311EA 454 EUR
ABNOMAB11311
Anti-KRT1 Mouse Monoclonal Antibody [clone: AE1 + AE3]
Antikörper
Mouse monoclonal antibody raised against Pan Cytokeratin.

Recommended Dilutions: Western Blot: 0.1 - 1.0 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:200 - 1:400

Type: Primary
Antigen: KRT1
Clonality: Monoclonal
Clone: AE1 + AE3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR