Anti-PTPRC Mouse Monoclonal Antibody [clone: 158-4D3]

Lieferant: Abnova
ABNOMAB11299EA 454 EUR
ABNOMAB11299
Anti-PTPRC Mouse Monoclonal Antibody [clone: 158-4D3]
Antikörper
Mouse monoclonal antibody raised against CD45RA (PTPRC).

  • Recommended Dilutions: Flow Cytometry: 1 μg/10^6 cells; Immunocytochemistry/Immunofluorescence: 1:10-1:2000; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:500; Immunoprecipitation: 1:10-1:500

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: 158-4D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR