Anti-BrdU Mouse Monoclonal Antibody [clone: BRD469]

Lieferant: Abnova
ABNOMAB11482EA 454 EUR
ABNOMAB11482
Anti-BrdU Mouse Monoclonal Antibody [clone: BRD469]
Antikörper
mouse monoclonal antibody raised against BrdU.

Recommended Dilutions: Flow Cytometry: 0.5-1 ?g/million cells; Immunofluorescence: 0.5-1 ?g/ml; Immunohistochemistry: 0.5-1 ?g/ml for 30 min at RT)

Type: Primary
Antigen: BrdU
Clonality: Monoclonal
Clone: BRD469
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR