Anti-HSPA1A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 5A5]

Lieferant: Abnova
ABNOMAB11400EA 643 EUR
ABNOMAB11400
Anti-HSPA1A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 5A5]
Antikörper
Mouse monoclonal antibody raised against recombinant HSPA1A.

Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:500; Immunoprecipitation: 1 ?g

Type: Primary
Antigen: HSPA1A
Clonality: Monoclonal
Clone: 5A5
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Amphibian, Chicken, Fish, Drosophila, Human, Mouse, Rat, Yeast
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR