Anti-DUX4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: P2B1]

Lieferant: Abnova
ABNOMAB11452EA 568 EUR
ABNOMAB11452
Anti-DUX4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: P2B1]
Antikörper
Mouse monoclonal antibody raised against recombinant partial DUX4.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: DUX4
Clonality: Monoclonal
Clone: P2B1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR