Anti-MDC1 Mouse Monoclonal Antibody (ATTO 655) [clone: P2B11]

Lieferant: Abnova
ABNOMAB11466EA 568 EUR
ABNOMAB11466
Anti-MDC1 Mouse Monoclonal Antibody (ATTO 655) [clone: P2B11]
Mouse monoclonal antibody raised against recombinant partial Mdc1.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: MDC1
Clonality: Monoclonal
Clone: P2B11
Conjugation: ATTO 655
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Chimpanzee, Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR