Anti-MCAM Mouse Monoclonal Antibody [clone: OJ79c]

Lieferant: Abnova
ABNOMAB11364EA 454 EUR
ABNOMAB11364
Anti-MCAM Mouse Monoclonal Antibody [clone: OJ79c]
Antikörper
Mouse monoclonal antibody raised against recombinant MCAM.

Recommended Dilutions: Flow Cytometry: 0.5-1 ?g/million cells; Immunofluorescence: 0.5-1 ?g/ml; Immunohistochemistry (Frozen sections): 0.5-1 ?g/ml for 30 min at RT; Immunoprecipitation: 1-2 ?g/500 ?g protein lysate; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: MCAM
Clonality: Monoclonal
Clone: OJ79c
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR