Anti-GFAP Mouse Monoclonal Antibody [clone: GA5]

Lieferant: Abnova
ABNOMAB11287EA 454 EUR
ABNOMAB11287
Anti-GFAP Mouse Monoclonal Antibody [clone: GA5]
Antikörper
Mouse monoclonal antibody raised against GFAP.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:200-1:400; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: GA5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR