Anti-EV71 VP1 Mouse Polyclonal Antibody
Lieferant: Abnova
ABNOPAB7630-B01PEA 406 EUR
ABNOPAB7630-B01P
Anti-EV71 VP1 Mouse Polyclonal Antibody
Antikörper
Mouse polyclonal antibody raised against EV71 VP1 protein.
Type: Primary
Antigen: EV71 VP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestellen