Anti-TGM4 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25811EA 568 EUR
ABNOPAB25811
Anti-TGM4 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant TGM4.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:200-1:500

Type: Primary
Antigen: TGM4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR