Anti-NKX2-4 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25799EA 454 EUR
ABNOPAB25799
Anti-NKX2-4 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of NKX2-4.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 2.5 ?g/ml

Type: Primary
Antigen: NKX2-4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR