Anti-GBP6 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25790EA 454 EUR
ABNOPAB25790
Anti-GBP6 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of GBP6.

Recommended Dilutions: Western Blot: 0.5 ?g/ml; Immunocytochemistry: 2.5 ?g/ml

Type: Primary
Antigen: GBP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR