Anti-ANKLE2 Chicken Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25771EA 518 EUR
ABNOPAB25771
Anti-ANKLE2 Chicken Polyclonal Antibody
Antikörper
Chicken polyclonal antibody raised against synthetic peptide of ANKLE2.

Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: ANKLE2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR