Anti-SLC35D2 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25738EA 454 EUR
ABNOPAB25738
Anti-SLC35D2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of SLC35D2.

Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunocytochemistry: 5 ?g/ml

Type: Primary
Antigen: SLC35D2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR