Anti-NETO1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25721EA 518 EUR
ABNOPAB25721
Anti-NETO1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of NETO1.

Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry: 2.5 ?g/ml

Type: Primary
Antigen: NETO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR