Anti-TFEB Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25696EA 454 EUR
ABNOPAB25696
Anti-TFEB Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of TFEB.

  • Recommended Dilutions: Western Blot: 1-2 μg/ml; Immunohistochemistry: 2.5 μg/ml

Type: Primary
Antigen: TFEB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR