Anti-ZMYM4 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25659EA 454 EUR
ABNOPAB25659
Anti-ZMYM4 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of ZMYM4.

Recommended Dilutions: Immunohistochemistry: 5 ?g/ml

Type: Primary
Antigen: ZMYM4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR