Anti-TCTEX1D2 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25652EA 454 EUR
ABNOPAB25652
Anti-TCTEX1D2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of TCTEX1D2.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: TCTEX1D2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR