Anti-ERBB2 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25609EA 643 EUR
ABNOPAB25609
Anti-ERBB2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of ERBB2.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 5-10 ?g/ml

Type: Primary
Antigen: ERBB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Gorilla, Horse, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR