Anti-USP2 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25606EA 568 EUR
ABNOPAB25606
Anti-USP2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of USP2.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 5 ?g/ml

Type: Primary
Antigen: USP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Rabbit, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR