Anti-ADGRB1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25593EA 643 EUR
ABNOPAB25593
Anti-ADGRB1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of BAI1.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 64 ?g/ml

Type: Primary
Antigen: ADGRB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR