Anti-ADCYAP1R1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25588EA 568 EUR
ABNOPAB25588
Anti-ADCYAP1R1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of ADCYAP1R1.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 7 ?g/ml

Type: Primary
Antigen: ADCYAP1R1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR