Anti-HTR2A Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25542EA 568 EUR
ABNOPAB25542
Anti-HTR2A Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of HTR2A.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 10-12.5 ?g/ml

Type: Primary
Antigen: HTR2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Gorilla, Horse, Human, Orangutan, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR