Anti-ADRB2 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25515EA 568 EUR
ABNOPAB25515
Anti-ADRB2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of ADRB2.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 5 ?g/ml

Type: Primary
Antigen: ADRB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Gorilla, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR